COM638472056 Combi-Blocks, Inc.

Methyl (2s)-2-amino-3,3-dimethylbutanoate

  • MDL number MFCD00137409
  • Cas Number : 63038-26-6
  • Molecular Weight : 145.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0236

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0236
Availability details for
Quantity
  • Availability
  • Add to Cart