COM945365210 Combi-Blocks, Inc.

3,5-Dibromo-4-chloroaniline

  • MDL number MFCD18914595
  • Cas Number : 35754-04-2
  • Molecular Weight : 285.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3294

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3294
Availability details for
Quantity
  • Availability
  • Add to Cart