COM945365211 Combi-Blocks, Inc.

3-(Aminomethyl)-6-(trifluoromethyl)pyridine HCl

  • MDL number MFCD26517257
  • Cas Number : 1956369-51-9
  • Molecular Weight : 212.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3465

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3465
Availability details for
Quantity
  • Availability
  • Add to Cart