COM945365218 Combi-Blocks, Inc.

(2-chloro-4,5-dimethylphenyl)boronic acid

  • MDL number MFCD24217817
  • Cas Number : 2246683-46-3
  • Molecular Weight : 184.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1326

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1326
Availability details for
Quantity
  • Availability
  • Add to Cart