COM945365219 Combi-Blocks, Inc.

4-chloro-5-(dihydroxyboranyl)benzene-1,2-dicarboxylic acid

  • MDL number MFCD30726946
  • Molecular Weight : 244.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1327

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1327
Availability details for
Quantity
  • Availability
  • Add to Cart