COM945365220 Combi-Blocks, Inc.

{3-[5-(ethoxycarbonyl)-1,3-thiazol-2-yl]phenyl}boronic acid

  • MDL number MFCD30726947
  • Molecular Weight : 277.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1332

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1332
Availability details for
Quantity
  • Availability
  • Add to Cart