COM945365221 Combi-Blocks, Inc.

2-[3-(dihydroxyboranyl)phenyl]-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD30726948
  • Molecular Weight : 249.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1333

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1333
Availability details for
Quantity
  • Availability
  • Add to Cart