COM945365222 Combi-Blocks, Inc.

[2-(benzyloxy)-3-cyano-5-fluorophenyl]boronic acid

  • MDL number MFCD30726949
  • Molecular Weight : 271.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1340

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1340
Availability details for
Quantity
  • Availability
  • Add to Cart