COM945365223 Combi-Blocks, Inc.

[4-fluoro-5-nitro-2-(trifluoromethyl)phenyl]boronic acid

  • MDL number MFCD30726950
  • Molecular Weight : 252.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1352

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1352
Availability details for
Quantity
  • Availability
  • Add to Cart