COM945365224 Combi-Blocks, Inc.

[3-(4-phenyl-1,3-thiazol-2-yl)phenyl]boronic acid

  • MDL number MFCD30726951
  • Molecular Weight : 281.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1366

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1366
Availability details for
Quantity
  • Availability
  • Add to Cart