COM945365232 Combi-Blocks, Inc.

2-Chloro-3-methoxypyridine-4-boronic acid

  • MDL number MFCD16610616
  • Cas Number : 957060-88-7
  • Molecular Weight : 187.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FB-0772

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FB-0772
Availability details for
Quantity
  • Availability
  • Add to Cart