COM945365236 Combi-Blocks, Inc.

5-Chloro-4-methoxycarbonyl-2-fluorophenylboronic acid pinacol ester

  • MDL number MFCD16996259
  • Cas Number : 890839-31-3
  • Molecular Weight : 314.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FF-7765

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FF-7765
Availability details for
Quantity
  • Availability
  • Add to Cart