COM945365238 Combi-Blocks, Inc.

5-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole, HCl

  • MDL number MFCD30726954
  • Cas Number : 2377608-65-4
  • Molecular Weight : 280.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2186

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2186
Availability details for
Quantity
  • Availability
  • Add to Cart