COM945365239 Combi-Blocks, Inc.

2-Methyl-1H-indole-7-boronic acid pinacol ester

  • MDL number MFCD09027760
  • Cas Number : 919119-59-8
  • Molecular Weight : 257.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2773

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2773
Availability details for
Quantity
  • Availability
  • Add to Cart