COM945365241 Combi-Blocks, Inc.

4-(Pyrazol-1-yl)piperidine hydrochloride

  • MDL number MFCD15072073
  • Cas Number : 690261-87-1
  • Molecular Weight : 187.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4257

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4257
Availability details for
Quantity
  • Availability
  • Add to Cart