COM945365242 Combi-Blocks, Inc.

2-Methoxy-3-nitrobenzamide

  • MDL number MFCD18398355
  • Cas Number : 722538-98-9
  • Molecular Weight : 196.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4403

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4403
Availability details for
Quantity
  • Availability
  • Add to Cart