COM945365246 Combi-Blocks, Inc.

tert-Butyl(3-fluorophenoxy)dimethylsilane

  • MDL number MFCD30296256
  • Cas Number : 114635-97-1
  • Molecular Weight : 226.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4430

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4430
Availability details for
Quantity
  • Availability
  • Add to Cart