COM945365247 Combi-Blocks, Inc.

6-bromo-1-methylpyrazolo[3,4-b]pyridine

  • MDL number MFCD22398843
  • Cas Number : 1289261-71-7
  • Molecular Weight : 212.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-1852

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HI-1852
Availability details for
Quantity
  • Availability
  • Add to Cart