COM945365248 Combi-Blocks, Inc.

2H-pyrazol-3-ylmethanol HCl

  • MDL number MFCD28392202
  • Cas Number : 270920-41-7
  • Molecular Weight : 134.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-1974

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HI-1974
Availability details for
Quantity
  • Availability
  • Add to Cart