COM945365257 Combi-Blocks, Inc.

methyl 4,5-diamino-2-chlorobenzoate

  • MDL number MFCD17168930
  • Cas Number : 64617-64-7
  • Molecular Weight : 200.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1495

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1495
Availability details for
Quantity
  • Availability
  • Add to Cart