COM945365258 Combi-Blocks, Inc.

1-phenylpyridin-2(1H)-one

  • MDL number MFCD01859711
  • Cas Number : 13131-02-7
  • Molecular Weight : 171.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1496

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1496
Availability details for
Quantity
  • Availability
  • Add to Cart