COM945365263 Combi-Blocks, Inc.

tert-butyl N-(2-bromo-4-chlorophenyl)carbamate

  • MDL number MFCD11975574
  • Cas Number : 384793-16-2
  • Molecular Weight : 306.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1519

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1519
Availability details for
Quantity
  • Availability
  • Add to Cart