COM945365268 Combi-Blocks, Inc.

tert-butyl N-(5-chloro-3-iodopyridin-2-yl)carbamate

  • MDL number MFCD30726959
  • Cas Number : 2092000-52-5
  • Molecular Weight : 354.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1564

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1564
Availability details for
Quantity
  • Availability
  • Add to Cart