COM945365271 Combi-Blocks, Inc.

8-fluoroquinoline-5-carbaldehyde

  • MDL number MFCD29761560
  • Cas Number : 1936339-14-8
  • Molecular Weight : 175.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1572

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1572
Availability details for
Quantity
  • Availability
  • Add to Cart