COM945365274 Combi-Blocks, Inc.

2-methoxyquinolin-6-ol

  • MDL number MFCD18414690
  • Cas Number : 623147-03-5
  • Molecular Weight : 175.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1590

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1590
Availability details for
Quantity
  • Availability
  • Add to Cart