COM945365275 Combi-Blocks, Inc.

2,6-dimethoxyquinoline

  • MDL number MFCD28337067
  • Cas Number : 222317-31-9
  • Molecular Weight : 189.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1591

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1591
Availability details for
Quantity
  • Availability
  • Add to Cart