COM945365278 Combi-Blocks, Inc.

3-bromo-1,4-difluoro-2-methyl-5-nitrobenzene

  • MDL number MFCD30726963
  • Cas Number : 2092001-02-8
  • Molecular Weight : 252.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1600

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1600
Availability details for
Quantity
  • Availability
  • Add to Cart