COM945365279 Combi-Blocks, Inc.

3-bromo-2,5-difluoro-4-methylaniline

  • MDL number MFCD30726964
  • Cas Number : 2092001-78-8
  • Molecular Weight : 222.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1601

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1601
Availability details for
Quantity
  • Availability
  • Add to Cart