COM945365281 Combi-Blocks, Inc.

8-hydroxyquinoline-4-carbonitrile

  • MDL number MFCD18415950
  • Cas Number : 1025556-11-9
  • Molecular Weight : 170.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1603

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1603
Availability details for
Quantity
  • Availability
  • Add to Cart