COM945365288 Combi-Blocks, Inc.

methyl 3-chloropyridazine-4-carboxylate

  • MDL number MFCD30726965
  • Cas Number : 1895477-24-3
  • Molecular Weight : 172.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1653

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1653
Availability details for
Quantity
  • Availability
  • Add to Cart