COM945365293 Combi-Blocks, Inc.

5-bromo-2-ethoxy-3-(trifluoromethyl)pyridine

  • MDL number MFCD28675388
  • Cas Number : 849934-83-4
  • Molecular Weight : 270.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1658

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1658
Availability details for
Quantity
  • Availability
  • Add to Cart