COM945365296 Combi-Blocks, Inc.

2-(benzyloxy)-5-bromo-3-(trifluoromethyl)pyridine

  • MDL number MFCD17168431
  • Cas Number : 1824262-21-6
  • Molecular Weight : 332.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1661

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1661
Availability details for
Quantity
  • Availability
  • Add to Cart