COM945365297 Combi-Blocks, Inc.

5-bromo-2-(2-methoxyethoxy)-3-(trifluoromethyl)pyridine

  • MDL number MFCD30726970
  • Cas Number : 2092000-72-9
  • Molecular Weight : 300.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1662

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1662
Availability details for
Quantity
  • Availability
  • Add to Cart