COM945365299 Combi-Blocks, Inc.

2-(benzyloxy)-3-methoxyaniline

  • MDL number MFCD30726972
  • Cas Number : 70978-05-1
  • Molecular Weight : 229.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1664

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1664
Availability details for
Quantity
  • Availability
  • Add to Cart