COM945365301 Combi-Blocks, Inc.

tert-butyl N-(3-bromo-5-cyanophenyl)carbamate

  • MDL number MFCD30726975
  • Cas Number : 2092002-01-0
  • Molecular Weight : 297.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1676

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1676
Availability details for
Quantity
  • Availability
  • Add to Cart