COM945365302 Combi-Blocks, Inc.

2-Iodo-3-methyl-5-nitropyridine

  • MDL number MFCD28666728
  • Cas Number : 49767-15-9
  • Molecular Weight : 264.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1680

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1680
Availability details for
Quantity
  • Availability
  • Add to Cart