COM945365304 Combi-Blocks, Inc.

1-(4-Cyanophenyl)-N-cyclohexylmethanesulfonamide

  • MDL number MFCD12531131
  • Cas Number : 1036520-78-1
  • Molecular Weight : 278.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1690

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1690
Availability details for
Quantity
  • Availability
  • Add to Cart