COM945365310 Combi-Blocks, Inc.

tert-butyl N-[(tert-butoxy)carbonyl]-N-methanesulfonylcarbamate

  • MDL number MFCD30726977
  • Cas Number : 2092000-74-1
  • Molecular Weight : 295.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1709

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1709
Availability details for
Quantity
  • Availability
  • Add to Cart