COM945365313 Combi-Blocks, Inc.

1-(2-Cyanophenyl)-N,N-dimethylmethanesulfonamide

  • MDL number MFCD11114768
  • Cas Number : 1036501-57-1
  • Molecular Weight : 224.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1713

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1713
Availability details for
Quantity
  • Availability
  • Add to Cart