COM945365314 Combi-Blocks, Inc.

1-(2-Cyanophenyl)-N-cyclopropylmethanesulfonamide

  • MDL number MFCD11123346
  • Cas Number : 1040339-14-7
  • Molecular Weight : 236.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1714

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1714
Availability details for
Quantity
  • Availability
  • Add to Cart