COM945365315 Combi-Blocks, Inc.

1-(2-Cyanophenyl)-N-isopropylmethanesulfonamide

  • MDL number MFCD07852063
  • Cas Number : 31846-43-2
  • Molecular Weight : 238.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1715

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1715
Availability details for
Quantity
  • Availability
  • Add to Cart