COM945365316 Combi-Blocks, Inc.

N-Benzyl-1-(2-cyanophenyl)methanesulfonamide

  • MDL number MFCD13353241
  • Cas Number : 31846-44-3
  • Molecular Weight : 286.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1716

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1716
Availability details for
Quantity
  • Availability
  • Add to Cart