COM945365322 Combi-Blocks, Inc.

1-(4-Cyanophenyl)-N-phenylmethanesulfonamide

  • MDL number MFCD12737917
  • Cas Number : 1040026-51-4
  • Molecular Weight : 272.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1723

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1723
Availability details for
Quantity
  • Availability
  • Add to Cart