COM945365323 Combi-Blocks, Inc.

4-[(Morpholine-4-sulfonyl)methyl]benzonitrile

  • MDL number MFCD11122872
  • Cas Number : 1042507-00-5
  • Molecular Weight : 266.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1724

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1724
Availability details for
Quantity
  • Availability
  • Add to Cart