COM945365327 Combi-Blocks, Inc.

tert-butyl 3-(benzyloxy)-2-{[(tert-butoxy)carbonyl]amino}propanoate

  • MDL number MFCD30726978
  • Cas Number : 95319-02-1
  • Molecular Weight : 351.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1731

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1731
Availability details for
Quantity
  • Availability
  • Add to Cart