COM945365328 Combi-Blocks, Inc.

ethyl 1H-imidazo[1,2-b]pyrazole-5-carboxylate

  • MDL number MFCD20257240
  • Cas Number : 1378954-67-6
  • Molecular Weight : 179.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1732

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1732
Availability details for
Quantity
  • Availability
  • Add to Cart