COM945365329 Combi-Blocks, Inc.

5-Bromo-6-chloro-N,N-dimethylpyridin-2-amine

  • MDL number MFCD26516998
  • Cas Number : 1823889-07-1
  • Molecular Weight : 235.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1882

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1882
Availability details for
Quantity
  • Availability
  • Add to Cart