COM945365334 Combi-Blocks, Inc.

BOC-Glu(OBzl)-Gly-OH

  • MDL number MFCD30726979
  • Cas Number : 51782-82-2
  • Molecular Weight : 394.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2579

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2579
Availability details for
Quantity
  • Availability
  • Add to Cart