COM945365335 Combi-Blocks, Inc.

N-(2,5-Dibromophenyl)-2,2,2-trifluoroacetamide

  • MDL number MFCD21415835
  • Cas Number : 1494061-58-3
  • Molecular Weight : 346.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4266

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4266
Availability details for
Quantity
  • Availability
  • Add to Cart