COM945365336 Combi-Blocks, Inc.

tert-Butyl (3S)-3-({[(4-methylbenzene)sulfonyl]oxy}methyl)pyrrolidine-1-carboxylate

  • MDL number MFCD22681619
  • Cas Number : 1391730-27-0
  • Molecular Weight : 355.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4301

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4301
Availability details for
Quantity
  • Availability
  • Add to Cart